Learn More
Meso Scale Discovery RPLEX HUM Heme Oxygenase 1 ASY
SDP
Catalog No. 5000078218
Encompass
The R-PLEX Human Heme Oxygenase 1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Heme Oxygenase 1 (HO-1) in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator This assay allows for the quantitative measurement of HO-1 levels with high precision and reproducibility
Catalog No. 50-000-78218 Supplier Meso Scale Discovery Supplier No. K151R8R2
The R-PLEX Human Heme Oxygenase 1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Heme Oxygenase 1 (HO-1) in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator This assay allows for the quantitative measurement of HO-1 levels with high precision and reproducibility