Learn More
Meso Scale Discovery RPLEX HUM Heme Oxygenase ASYQP
SDP
Catalog No. 5000078261
Encompass
The R-PLEX Human Heme Oxygenase 1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Heme Oxygenase 1 (HO-1) in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator It can test up to 80 samples in duplicate and only requires a small sample volume of 50 L The assay takes around 3 5 hours to complete
Catalog No. 50-000-78261 Supplier Meso Scale Discovery Supplier No. K151R8R22
The R-PLEX Human Heme Oxygenase 1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Heme Oxygenase 1 (HO-1) in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator It can test up to 80 samples in duplicate and only requires a small sample volume of 50 L The assay takes around 3 5 hours to complete