Learn More
Meso Scale Discovery RPLEX HUM IA2/RPTPN ASYQP
SDP
Catalog No. 5000080153
Encompass
The Meso Scale Discovery R-PLEX Human IA-2 (RPTP-N) Assay is a multiplex immunoassay panel that allows simultaneous measurement of IA-2 autoantibodies in human serum plasma and other biological matrices This assay utilizes electrochemiluminescent detection technology on patterned arrays for high sensitivity wide dynamic range and low sample volume requirements The panel provides a quantitative read-out of IA-2 autoantibody concentrations to aid in the diagnosis and monitoring of type 1 diabetes This assay requires the Meso Scale Discovery SECTOR Imager and provides a rapid reproducible method to profile IA-2 autoantibodies
Catalog No. 50-000-80153 Supplier Meso Scale Discovery Supplier No. K151AKAR22
The Meso Scale Discovery R-PLEX Human IA-2 (RPTP-N) Assay is a multiplex immunoassay panel that allows simultaneous measurement of IA-2 autoantibodies in human serum plasma and other biological matrices This assay utilizes electrochemiluminescent detection technology on patterned arrays for high sensitivity wide dynamic range and low sample volume requirements The panel provides a quantitative read-out of IA-2 autoantibody concentrations to aid in the diagnosis and monitoring of type 1 diabetes This assay requires the Meso Scale Discovery SECTOR Imager and provides a rapid reproducible method to profile IA-2 autoantibodies