Learn More
Meso Scale Discovery RPLEX HUM IGF2R/CIM6PR ASY
SDP
Catalog No. 5000079649
Encompass
The Meso Scale Discovery IGF-2R (CI-MPR) Kit is an electrochemiluminescent immunoassay designed for the quantitative measurement of Insulin-like Growth Factor 2 Receptor (IGF-2R) in human serum plasma and cell culture supernatants This assay employs an MSD GOLD SULFO-TAG label and has a typical lower limit of detection of 0 03 ng/mL The kit includes a 96-well plate coated with an IGF-2R capture antibody a detection antibody conjugated to an MSD SULFO-TAG calibrators positive and negative controls 10X Read Buffer 10X Wash Buffer and Blocker A This assay can be run on the MESO QuickPlex SQ 120 MESO SECTOR S 600 or MESO SECTOR S 6000 instruments
Catalog No. 50-000-79649 Supplier Meso Scale Discovery Supplier No. K151C4R2
The Meso Scale Discovery IGF-2R (CI-MPR) Kit is an electrochemiluminescent immunoassay designed for the quantitative measurement of Insulin-like Growth Factor 2 Receptor (IGF-2R) in human serum plasma and cell culture supernatants This assay employs an MSD GOLD SULFO-TAG label and has a typical lower limit of detection of 0 03 ng/mL The kit includes a 96-well plate coated with an IGF-2R capture antibody a detection antibody conjugated to an MSD SULFO-TAG calibrators positive and negative controls 10X Read Buffer 10X Wash Buffer and Blocker A This assay can be run on the MESO QuickPlex SQ 120 MESO SECTOR S 600 or MESO SECTOR S 6000 instruments