Learn More
Meso Scale Discovery RPLEX HUM IL28BIFN 3 ASY
SDP
Catalog No. 5000079180
Encompass
The R-PLEX Human IL-28B (IFN-lambda3) Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that enables the quantitative measurement of human interleukin 28B (IL-28B) also known as interferon lambda 3 (IFN-lambda3) This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The assay has a detection range of 0 03-1000 pg/mL and requires 50 L of sample per well This product can be used to measure IL-28B levels in serum plasma and cell culture supernatants IL-28B is a cytokine that has antiviral activity and is involved in immune response against viruses
Catalog No. 50-000-79180 Supplier Meso Scale Discovery Supplier No. K151Q4R2
The R-PLEX Human IL-28B (IFN-lambda3) Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that enables the quantitative measurement of human interleukin 28B (IL-28B) also known as interferon lambda 3 (IFN-lambda3) This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The assay has a detection range of 0 03-1000 pg/mL and requires 50 L of sample per well This product can be used to measure IL-28B levels in serum plasma and cell culture supernatants IL-28B is a cytokine that has antiviral activity and is involved in immune response against viruses