Learn More
Meso Scale Discovery RPLEX HUM IL28BIFN 3 ASYQP
SDP
Catalog No. 5000078576
Encompass
The R-PLEX Human IL-28B (IFN Lambda3) Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures the cytokine IL-28B (IFN Lambda3) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is designed for use on MSD s QuickPlex SQ 120 instrument The assay has a detection range of 0 06-1500 pg/mL and requires 25 L or less of sample The total assay time is approximately 3 5 hours This assay provides a sensitive accurate method for quantifying IL-28B levels in biological samples
Catalog No. 50-000-78576 Supplier Meso Scale Discovery Supplier No. K151Q4R22
The R-PLEX Human IL-28B (IFN Lambda3) Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures the cytokine IL-28B (IFN Lambda3) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is designed for use on MSD s QuickPlex SQ 120 instrument The assay has a detection range of 0 06-1500 pg/mL and requires 25 L or less of sample The total assay time is approximately 3 5 hours This assay provides a sensitive accurate method for quantifying IL-28B levels in biological samples