Learn More
Meso Scale Discovery RPLEX HUM Influenza A NP ASY
SDP
Catalog No. 5000078915
Encompass
The R-PLEX Human Influenza A NP Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that simultaneously detects influenza A nucleoprotein and differentiates between H1N1 and H3N2 subtypes in human nasopharyngeal swab specimens This assay utilizes MSD s electrochemiluminescent detection technology and provides results in under 5 hours It is intended for research use only and not for use in diagnostic procedures The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer blocker solutions calibrators and controls
Catalog No. 50-000-78915 Supplier Meso Scale Discovery Supplier No. K1519GR2
The R-PLEX Human Influenza A NP Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that simultaneously detects influenza A nucleoprotein and differentiates between H1N1 and H3N2 subtypes in human nasopharyngeal swab specimens This assay utilizes MSD s electrochemiluminescent detection technology and provides results in under 5 hours It is intended for research use only and not for use in diagnostic procedures The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer blocker solutions calibrators and controls