Learn More
Meso Scale Discovery RPLEX HUM Influenza A NP ASYQP
SDP
Catalog No. 5000080327
Encompass
The R-PLEX Human Influenza A NP Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that simultaneously detects influenza A nucleoprotein and differentiates between H1 H3 and H5 subtypes in human nasal swab samples This assay utilizes electrochemiluminescence detection technology and provides results in under 3 hours With high sensitivity and specificity it can aid in the rapid diagnosis of influenza A infections The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls assay and wash buffers substrate and plate seals sufficient for one 96-well plate
Catalog No. 50-000-80327 Supplier Meso Scale Discovery Supplier No. K1519GR22
The R-PLEX Human Influenza A NP Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that simultaneously detects influenza A nucleoprotein and differentiates between H1 H3 and H5 subtypes in human nasal swab samples This assay utilizes electrochemiluminescence detection technology and provides results in under 3 hours With high sensitivity and specificity it can aid in the rapid diagnosis of influenza A infections The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls assay and wash buffers substrate and plate seals sufficient for one 96-well plate