Learn More
Meso Scale Discovery RPLEX HUM Kallikrein3/PSA ASY
SDP
Catalog No. 5000078917
Encompass
The R-PLEX Human Kallikrein 3 (PSA) Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of total prostate-specific antigen (PSA) in human serum plasma and urine This assay utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a broad dynamic range The assay has a lower limit of detection of 0 01 ng/mL PSA and an upper limit of 100 ng/mL PSA The assay requires 25 L of sample and can be completed in under 3 hours This assay provides a sensitive accurate method for measuring PSA levels
Catalog No. 50-000-78917 Supplier Meso Scale Discovery Supplier No. K151ACJR2
The R-PLEX Human Kallikrein 3 (PSA) Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of total prostate-specific antigen (PSA) in human serum plasma and urine This assay utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a broad dynamic range The assay has a lower limit of detection of 0 01 ng/mL PSA and an upper limit of 100 ng/mL PSA The assay requires 25 L of sample and can be completed in under 3 hours This assay provides a sensitive accurate method for measuring PSA levels