Learn More
Meso Scale Discovery RPLEX HUM Lactotransferrin ASY
SDP
Catalog No. 5000081582
Encompass
The Meso Scale Discovery R-PLEX Human Lactotransferrin Assay QuickPlex 5-PL Kit is a multiplex immunoassay that quantitatively measures lactotransferrin in human serum plasma saliva milk and cell culture supernatant samples The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 5-plex kit includes a plate coated with capture antibodies detection antibodies calibrators controls assay and wash buffers substrate and plastic consumables needed to run the assay The kit can test up to 80 samples in duplicate and has a testing time of approximately 3 5 hours
Catalog No. 50-000-81582 Supplier Meso Scale Discovery Supplier No. K151ALAR2
The Meso Scale Discovery R-PLEX Human Lactotransferrin Assay QuickPlex 5-PL Kit is a multiplex immunoassay that quantitatively measures lactotransferrin in human serum plasma saliva milk and cell culture supernatant samples The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 5-plex kit includes a plate coated with capture antibodies detection antibodies calibrators controls assay and wash buffers substrate and plastic consumables needed to run the assay The kit can test up to 80 samples in duplicate and has a testing time of approximately 3 5 hours