Learn More
Meso Scale Discovery RPLEX HUM LDL Receptor ASYQP
SDP
Catalog No. 5000079424
Encompass
The R-PLEX Human LDL Receptor Assay from Meso Scale Discovery is a multiplex immunoassay that measures the LDL receptor in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to quantitatively measure LDL receptor levels This assay allows for the rapid accurate analysis of LDL receptor levels which can provide insight into cholesterol metabolism and cardiovascular disease risk
Catalog No. 50-000-79424 Supplier Meso Scale Discovery Supplier No. K151ACWR22
The R-PLEX Human LDL Receptor Assay from Meso Scale Discovery is a multiplex immunoassay that measures the LDL receptor in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to quantitatively measure LDL receptor levels This assay allows for the rapid accurate analysis of LDL receptor levels which can provide insight into cholesterol metabolism and cardiovascular disease risk