Learn More
Meso Scale Discovery RPLEX HUM LRRK2 pS935 ASYQP
SDP
Catalog No. 5000080608
Encompass
The R-Plex Human LRRK2 pS935 Assay Kit from Meso Scale Diagnostics measures LRRK2 phosphorylated at serine 935 in human samples using electrochemiluminescence The assay uses a plate pre-coated with a capture antibody for total LRRK2 Samples bind to this antibody and a detection antibody for phospho-LRRK2 is added forming a sandwich complex A read buffer generates a signal proportional to phospho-LRRK2 levels The kit includes a plate detection antibody read buffer sample diluent and lyophilized calibrator
Catalog No. 50-000-80608 Supplier Meso Scale Discovery Supplier No. K1511QR22
The R-Plex Human LRRK2 pS935 Assay Kit from Meso Scale Diagnostics measures LRRK2 phosphorylated at serine 935 in human samples using electrochemiluminescence The assay uses a plate pre-coated with a capture antibody for total LRRK2 Samples bind to this antibody and a detection antibody for phospho-LRRK2 is added forming a sandwich complex A read buffer generates a signal proportional to phospho-LRRK2 levels The kit includes a plate detection antibody read buffer sample diluent and lyophilized calibrator