Learn More
Meso Scale Discovery RPLEX HUM Mcl1/BAK Compl ASYQP
SDP
Catalog No. 5000080159
Encompass
The R-PLEX Human Mcl-1/Bak Complex Assay Kit from Meso Scale Discovery is used to quantitatively measure Mcl-1/Bak protein-protein interactions in cell or tissue lysates This electrochemiluminescent assay utilizes capture antibodies against Mcl-1 immobilized on carbon electrodes Bak is detected using a SULFO-TAG labeled anti-Bak detection antibody The kit includes a 96-well plate coated with anti-Mcl-1 capture antibody anti-Bak detection antibody assay buffer wash buffer read buffer cell lysis buffer and positive control lysate The assay can detect endogenous levels of Mcl-1/Bak complexes and be used to screen for inhibitors of the interaction
Catalog No. 50-000-80159 Supplier Meso Scale Discovery Supplier No. K1518BR22
The R-PLEX Human Mcl-1/Bak Complex Assay Kit from Meso Scale Discovery is used to quantitatively measure Mcl-1/Bak protein-protein interactions in cell or tissue lysates This electrochemiluminescent assay utilizes capture antibodies against Mcl-1 immobilized on carbon electrodes Bak is detected using a SULFO-TAG labeled anti-Bak detection antibody The kit includes a 96-well plate coated with anti-Mcl-1 capture antibody anti-Bak detection antibody assay buffer wash buffer read buffer cell lysis buffer and positive control lysate The assay can detect endogenous levels of Mcl-1/Bak complexes and be used to screen for inhibitors of the interaction