Learn More
Meso Scale Discovery RPLEX HUM Mcl1/BAK Complex ASY
SDP
Catalog No. 5000080262
Encompass
The MCL-1/BAK Complex DuoSet IC ELISA is an in vitro assay for the quantitative measurement of the MCL-1/BAK protein complex in cell and tissue lysates It contains a capture antibody specific for MCL-1 and a detection antibody specific for BAK This ELISA allows for the specific and sensitive detection of the endogenous levels of the MCL-1/BAK complex With a detection range of 0 156-10 ng/mL and high specificity it is a useful tool for studying the role of the MCL-1/BAK interaction in apoptosis regulation The assay takes approximately 4 hours to complete
Catalog No. 50-000-80262 Supplier Meso Scale Discovery Supplier No. K1518BR2
The MCL-1/BAK Complex DuoSet IC ELISA is an in vitro assay for the quantitative measurement of the MCL-1/BAK protein complex in cell and tissue lysates It contains a capture antibody specific for MCL-1 and a detection antibody specific for BAK This ELISA allows for the specific and sensitive detection of the endogenous levels of the MCL-1/BAK complex With a detection range of 0 156-10 ng/mL and high specificity it is a useful tool for studying the role of the MCL-1/BAK interaction in apoptosis regulation The assay takes approximately 4 hours to complete