Learn More
Meso Scale Discovery RPLEX HUM Mcl1/Bcl2L3 ASY
SDP
Supplier:  Meso Scale Discovery K1516XR2
Encompass
The R-PLEX Human Mcl1 Bcl2-L Bcl-xL Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that allows for the simultaneous quantitative measurement of Mcl1 Bcl2-L and Bcl-xL proteins in human serum plasma and cell lysates using electrochemiluminescence detection technology The assay utilizes specific capture antibodies coated on distinct spots on a multi-array plate and ruthenium-labeled detection antibodies to provide sensitive and accurate measurements of these anti-apoptotic proteins which play important roles in regulating apoptosis and cancer cell survival This multiplex kit allows efficient quantification of these related proteins in a single small sample volume
Catalog No. 50-000-78885