Learn More
Meso Scale Discovery RPLEX HUM Mcl1/Bcl2L3 ASYQP
SDP
Catalog No. 5000078909
Encompass
The Meso Scale Discovery Human IL-22 Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin-22 (IL-22) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody to human IL-22 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 11 pg/mL and a 7-log dynamic range from 0 11-10 000 pg/mL Sample incubation time is 2 hours
Catalog No. 50-000-78909 Supplier Meso Scale Discovery Supplier No. K1516XR22
The Meso Scale Discovery Human IL-22 Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin-22 (IL-22) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody to human IL-22 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 11 pg/mL and a 7-log dynamic range from 0 11-10 000 pg/mL Sample incubation time is 2 hours