Learn More
Meso Scale Discovery RPLEX HUM Mcl1/BIM Complex ASY
SDP
Catalog No. 5000081716
Encompass
The R-PLEX Human Mcl-1 Bim Complex Assay Kit from Meso Scale Discovery is used to quantitatively measure Mcl-1 and Bim protein interactions in cell or tissue lysates This electrochemiluminescent assay utilizes patterned arrays and MSD s proprietary SULFO-TAG labels for high sensitivity and low background The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer blocker solutions SULFO-TAG labeled detection antibodies and calibrators to generate a standard curve This assay allows for the quantitative analysis of Mcl-1/Bim interactions involved in the regulation of apoptosis
Catalog No. 50-000-81716 Supplier Meso Scale Discovery Supplier No. K1518AR2
The R-PLEX Human Mcl-1 Bim Complex Assay Kit from Meso Scale Discovery is used to quantitatively measure Mcl-1 and Bim protein interactions in cell or tissue lysates This electrochemiluminescent assay utilizes patterned arrays and MSD s proprietary SULFO-TAG labels for high sensitivity and low background The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer blocker solutions SULFO-TAG labeled detection antibodies and calibrators to generate a standard curve This assay allows for the quantitative analysis of Mcl-1/Bim interactions involved in the regulation of apoptosis