Learn More
Meso Scale Discovery RPLEX HUM Mcl1BIM Cmplx ASYQP
SDP
Catalog No. 5000078938
Encompass
The R-PLEX Human Mcl-1 Bim Complex Assay Kit from Meso Scale Discovery is used to quantitatively measure Mcl-1 and Bim protein interactions in cell or tissue lysates This electrochemiluminescent assay utilizes capture antibodies against Mcl-1 or Bim immobilized on multi-array plates After incubation with cell lysate detection antibodies and read buffer are added to produce light signals proportional to Mcl-1/Bim binding that are measured on an MSD instrument The kit includes a plate antibodies calibrator and buffer sufficient for 96 tests to profile protein interactions influencing apoptosis regulation
Catalog No. 50-000-78938 Supplier Meso Scale Discovery Supplier No. K1518AR22
The R-PLEX Human Mcl-1 Bim Complex Assay Kit from Meso Scale Discovery is used to quantitatively measure Mcl-1 and Bim protein interactions in cell or tissue lysates This electrochemiluminescent assay utilizes capture antibodies against Mcl-1 or Bim immobilized on multi-array plates After incubation with cell lysate detection antibodies and read buffer are added to produce light signals proportional to Mcl-1/Bim binding that are measured on an MSD instrument The kit includes a plate antibodies calibrator and buffer sufficient for 96 tests to profile protein interactions influencing apoptosis regulation