Learn More
Meso Scale Discovery RPLEX HUM MPIF1/CCL23 ASYQP
SDP
Catalog No. 5000082088
Encompass
The R-PLEX Human MIF-1 (CCL23) Assay is a multiplex immunoassay panel that measures MIF-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and patterned arrays This assay has high sensitivity a broad dynamic range and requires low sample volume It is available in a 96-well plate format and has a testing time of approximately 3 5 hours The panel is validated for various biological samples and is ideal for studying MIF-1 biology evaluating drug efficacy and biomarker discovery
Catalog No. 50-000-82088 Supplier Meso Scale Discovery Supplier No. K151G1R22
The R-PLEX Human MIF-1 (CCL23) Assay is a multiplex immunoassay panel that measures MIF-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and patterned arrays This assay has high sensitivity a broad dynamic range and requires low sample volume It is available in a 96-well plate format and has a testing time of approximately 3 5 hours The panel is validated for various biological samples and is ideal for studying MIF-1 biology evaluating drug efficacy and biomarker discovery