Learn More
Meso Scale Discovery RPLEX HUM Neurofilament ASYQP
SDP
Catalog No. 5000078382
Encompass
The Meso Scale Discovery R-PLEX Human Neurofilament H Assay measures neurofilament heavy chain (pNF-H) in human serum plasma and CSF using electrochemiluminescence (ECL) detection It provides quantitative pNF-H levels to evaluate axonal damage with high sensitivity and a broad dynamic range for healthy and disease samples Requiring a small sample volume it delivers results in under 3 hours on a standard plate reader aiding researchers in assessing pNF-H as a biomarker for neurodegenerative diseases
Catalog No. 50-000-78382 Supplier Meso Scale Discovery Supplier No. K1517YR22
The Meso Scale Discovery R-PLEX Human Neurofilament H Assay measures neurofilament heavy chain (pNF-H) in human serum plasma and CSF using electrochemiluminescence (ECL) detection It provides quantitative pNF-H levels to evaluate axonal damage with high sensitivity and a broad dynamic range for healthy and disease samples Requiring a small sample volume it delivers results in under 3 hours on a standard plate reader aiding researchers in assessing pNF-H as a biomarker for neurodegenerative diseases