Learn More
Meso Scale Discovery RPLEX HUM Neuropilin1 ASYQP
SDP
Catalog No. 5000080325
Encompass
The R-PLEX Human Neuropilin-1 Assay is a multiplex immunoassay kit for measuring Neuropilin-1 in human serum plasma and cell culture supernatants It uses a quantitative sandwich enzyme immunoassay with antibodies pre-coated on 96-well plates The kit includes standards detection antibodies streptavidin-HRP buffers substrate and stop solution for colorimetric quantification It allows high-throughput screening with a sensitivity of 19 pg/mL providing a rapid sensitive and reproducible method to quantify Neuropilin-1 in various samples
Catalog No. 50-000-80325 Supplier Meso Scale Discovery Supplier No. K151ABWR22
The R-PLEX Human Neuropilin-1 Assay is a multiplex immunoassay kit for measuring Neuropilin-1 in human serum plasma and cell culture supernatants It uses a quantitative sandwich enzyme immunoassay with antibodies pre-coated on 96-well plates The kit includes standards detection antibodies streptavidin-HRP buffers substrate and stop solution for colorimetric quantification It allows high-throughput screening with a sensitivity of 19 pg/mL providing a rapid sensitive and reproducible method to quantify Neuropilin-1 in various samples