Learn More
Meso Scale Discovery RPLEX HUM Neurotrophin3 ASY
SDP
Catalog No. 5000078949
Encompass
The R-PLEX Human Neurotrophin-3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human neurotrophin-3 (NT-3) protein levels across samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies specific for NT-3 detection antibodies calibrators and reagents required to perform the assay Quantitative results can be obtained in as little as 3 5 hours This assay is for research use only
Catalog No. 50-000-78949 Supplier Meso Scale Discovery Supplier No. K1519VR2
The R-PLEX Human Neurotrophin-3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human neurotrophin-3 (NT-3) protein levels across samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies specific for NT-3 detection antibodies calibrators and reagents required to perform the assay Quantitative results can be obtained in as little as 3 5 hours This assay is for research use only