Learn More
Meso Scale Discovery RPLEX HUM Nucleotidase ASY
SDP
Catalog No. 5000080148
Encompass
The R-Plex Human 5 -Nucleotidase Assay from Meso Scale Diagnostics measures 5 -Nucleotidase in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) technology It offers high sensitivity a wide dynamic range and low sample volume requirements This assay quantifies 5 -Nucleotidase a glycoprotein in purine metabolism aiding in studying liver disease cancers and gout With a testing time of 3 5 hours it enables efficient analysis of 5 -Nucleotidase levels
Catalog No. 50-000-80148 Supplier Meso Scale Discovery Supplier No. K151AJFR2
The R-Plex Human 5 -Nucleotidase Assay from Meso Scale Diagnostics measures 5 -Nucleotidase in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) technology It offers high sensitivity a wide dynamic range and low sample volume requirements This assay quantifies 5 -Nucleotidase a glycoprotein in purine metabolism aiding in studying liver disease cancers and gout With a testing time of 3 5 hours it enables efficient analysis of 5 -Nucleotidase levels