Learn More
Meso Scale Discovery RPLEX HUM Osteoactivin ASYQP
SDP
Catalog No. 5000079866
Encompass
The Meso Scale Discovery R-PLEX Human Osteoactivin Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of osteoactivin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals It can accurately quantify osteoactivin levels and is suitable for studying bone metabolism osteoporosis and skeletal disorders
Catalog No. 50-000-79866 Supplier Meso Scale Discovery Supplier No. K151ZHR22
The Meso Scale Discovery R-PLEX Human Osteoactivin Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of osteoactivin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals It can accurately quantify osteoactivin levels and is suitable for studying bone metabolism osteoporosis and skeletal disorders