Learn More
Meso Scale Discovery RPLEX HUM Osteoprotegeri ASYQP
SDP
Catalog No. 5000081809
Encompass
The Meso Scale Discovery R-PLEX Human Osteoprotegerin Assay measures osteoprotegerin (OPG) in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection It offers high sensitivity a wide dynamic range and low sample volume needs This assay quantifies OPG crucial in bone remodeling by inhibiting osteoclasts It s useful in research on bone disorders like osteoporosis and cancer metastasis The kit includes a 96-well plate with capture antibody detection antibody read buffer calibrator and sample diluent
Catalog No. 50-000-81809 Supplier Meso Scale Discovery Supplier No. K151ZKR22
The Meso Scale Discovery R-PLEX Human Osteoprotegerin Assay measures osteoprotegerin (OPG) in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection It offers high sensitivity a wide dynamic range and low sample volume needs This assay quantifies OPG crucial in bone remodeling by inhibiting osteoclasts It s useful in research on bone disorders like osteoporosis and cancer metastasis The kit includes a 96-well plate with capture antibody detection antibody read buffer calibrator and sample diluent