Learn More
Meso Scale Discovery RPLEX HUM Osteoprotegerin ASY
SDP
Supplier:  Meso Scale Discovery K151ZKR2
Encompass
The Meso Scale Discovery R-PLEX Human Osteoprotegerin Assay is a multiplex immunoassay panel that measures osteoprotegerin (OPG) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms It is intended for research use only to study OPG levels and not for diagnostic procedures This assay measures total OPG and can aid in understanding bone remodeling processes
Catalog No. 50-000-78835