Learn More
Meso Scale Discovery RPLEX HUM p38 pT180/pY182 ASY
SDP
Catalog No. 5000081622
Encompass
The R-Plex Human p38 pT180/pY182 Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring phosphorylated p38 MAP kinase at Thr180 and Tyr182 in human cell lysates It uses electrochemiluminescent detection for rapid sensitive and reproducible analysis of p38 phosphorylation The kit includes a 96-well plate with capture antibodies detection antibodies and calibrator solutions It monitors p38 activation due to cellular stresses and inflammatory signals with a 2-hour sample incubation for high-throughput profiling
Catalog No. 50-000-81622 Supplier Meso Scale Discovery Supplier No. K151AKSR2
The R-Plex Human p38 pT180/pY182 Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring phosphorylated p38 MAP kinase at Thr180 and Tyr182 in human cell lysates It uses electrochemiluminescent detection for rapid sensitive and reproducible analysis of p38 phosphorylation The kit includes a 96-well plate with capture antibodies detection antibodies and calibrator solutions It monitors p38 activation due to cellular stresses and inflammatory signals with a 2-hour sample incubation for high-throughput profiling