Learn More
Meso Scale Discovery RPLEX HUM p38 pT180pY182 ASYQP
SDP
Catalog No. 5000081930
Encompass
The R-Plex Human p38 [pT180/pY182] Assay Kit from Meso Scale Discovery is used to quantitatively measure the levels of phosphorylated p38 MAPK at threonine 180 and tyrosine 182 in cell lysates This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide high sensitivity low background and a wide dynamic range The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium read buffer wash buffer blocker solutions calibrator and positive control lysate This assay allows for the rapid reproducible quantification of phospho-p38 levels for studying cellular signaling pathways
Catalog No. 50-000-81930 Supplier Meso Scale Discovery Supplier No. K151AKSR22
The R-Plex Human p38 [pT180/pY182] Assay Kit from Meso Scale Discovery is used to quantitatively measure the levels of phosphorylated p38 MAPK at threonine 180 and tyrosine 182 in cell lysates This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide high sensitivity low background and a wide dynamic range The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium read buffer wash buffer blocker solutions calibrator and positive control lysate This assay allows for the rapid reproducible quantification of phospho-p38 levels for studying cellular signaling pathways