Learn More
Meso Scale Discovery RPLEX HUM PDGFRB total ASY
SDP
Catalog No. 5000081426
Encompass
The R-PLEX Human PDGFR- Total Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the total levels of human platelet-derived growth factor receptor beta (PDGFR- ) in cell and tissue lysates using electrochemiluminescence detection technology This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and positive control lysate The assay has a detection range of 50-50 000 pg/mL and exhibits minimal cross-reactivity with other growth factors This product enables high-throughput quantification of total PDGFR- levels for research applications
Catalog No. 50-000-81426 Supplier Meso Scale Discovery Supplier No. K151W8R2
The R-PLEX Human PDGFR- Total Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the total levels of human platelet-derived growth factor receptor beta (PDGFR- ) in cell and tissue lysates using electrochemiluminescence detection technology This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and positive control lysate The assay has a detection range of 50-50 000 pg/mL and exhibits minimal cross-reactivity with other growth factors This product enables high-throughput quantification of total PDGFR- levels for research applications