Learn More
Meso Scale Discovery RPLEX HUM PRAS40 total ASY
SDP
Catalog No. 5000079611
Encompass
The R-PLEX Human PRAS40 Total Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of total PRAS40 protein in human cell and tissue samples The assay utilizes an electrochemiluminescent detection method and provides high sensitivity broad dynamic range and requires low sample input The kit includes a 96-well plate coated with capture antibody detection antibody read buffer sample diluent and vials of calibrator This assay allows for the simple and rapid analysis of PRAS40 protein levels which can provide insight into mTOR pathway biology and related disease states
Catalog No. 50-000-79611 Supplier Meso Scale Discovery Supplier No. K151AKUR2
Add to Cart
Add to Cart
The R-PLEX Human PRAS40 Total Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of total PRAS40 protein in human cell and tissue samples The assay utilizes an electrochemiluminescent detection method and provides high sensitivity broad dynamic range and requires low sample input The kit includes a 96-well plate coated with capture antibody detection antibody read buffer sample diluent and vials of calibrator This assay allows for the simple and rapid analysis of PRAS40 protein levels which can provide insight into mTOR pathway biology and related disease states