Learn More
Meso Scale Discovery RPLEX HUM PresenilinNTF ASYQP
SDP
Catalog No. 5000080358
Encompass
The R-Plex Human Presenilin 1 NTF Assay is a multiplex immunoassay kit for measuring the N-terminal fragment of Presenilin 1 in human samples like serum plasma and cell culture supernatants It uses a quantitative sandwich enzyme immunoassay and color-coded microspheres to measure Presenilin 1 NTF levels The kit includes microparticle cocktails standards controls diluents detection antibodies streptavidin-phycoerythrin wash solutions microplates seals and a protocol enabling high-throughput sensitive and reproducible quantification in research
Catalog No. 50-000-80358 Supplier Meso Scale Discovery Supplier No. K151ACLR22
The R-Plex Human Presenilin 1 NTF Assay is a multiplex immunoassay kit for measuring the N-terminal fragment of Presenilin 1 in human samples like serum plasma and cell culture supernatants It uses a quantitative sandwich enzyme immunoassay and color-coded microspheres to measure Presenilin 1 NTF levels The kit includes microparticle cocktails standards controls diluents detection antibodies streptavidin-phycoerythrin wash solutions microplates seals and a protocol enabling high-throughput sensitive and reproducible quantification in research