Learn More
Meso Scale Discovery RPLEX HUM Thrombomodulin ASYQP
SDP
Catalog No. 5000081843
Encompass
The R-Plex Human Thrombomodulin Assay from Meso Scale Discovery is a multiplex immunoassay that measures thrombomodulin levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements The assay can monitor thrombomodulin levels as a biomarker of endothelial cell activation and damage It is available as a singleplex or as a multiplex with other cardiovascular disease biomarkers The R-Plex assay delivers rapid results in just 3 5 hours
Catalog No. 50-000-81843 Supplier Meso Scale Discovery Supplier No. K151ZRR22
The R-Plex Human Thrombomodulin Assay from Meso Scale Discovery is a multiplex immunoassay that measures thrombomodulin levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements The assay can monitor thrombomodulin levels as a biomarker of endothelial cell activation and damage It is available as a singleplex or as a multiplex with other cardiovascular disease biomarkers The R-Plex assay delivers rapid results in just 3 5 hours