Learn More
Meso Scale Discovery RPLEX HUM TNFAIP6/TSG6 ASYQP
SDP
Catalog No. 5000078822
Encompass
The R-PLEX Human TNF IP6/TSG-6 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures TNF IP6/TSG-6 levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and a vial of lyophilized calibrator The assay has a typical sensitivity of 19 5 pg/mL and an upper limit of quantification of 50 000 pg/mL
Catalog No. 50-000-78822 Supplier Meso Scale Discovery Supplier No. K1510UR22
The R-PLEX Human TNF IP6/TSG-6 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures TNF IP6/TSG-6 levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and a vial of lyophilized calibrator The assay has a typical sensitivity of 19 5 pg/mL and an upper limit of quantification of 50 000 pg/mL