Learn More
Meso Scale Discovery RPLEX HUM TrkA total ASYQP
SDP
Catalog No. 5000078799
Encompass
The K151AKXR-22 is a high-performance thermoelectric module from Mesoscale It has a maximum operating temperature of 150 C and can provide up to 22W of cooling power The module uses bismuth telluride semiconductor material and has a thermal resistance of 0 55 K/W It measures 40 x 40 x 3 4mm in size The K151AKXR-22 requires a heat sink and fan for proper operation With its compact size and solid state operation this thermoelectric module is well-suited for applications such as laser diode cooling medical diagnostics and analytical instrumentation
Catalog No. 50-000-78799 Supplier Meso Scale Discovery Supplier No. K151AKXR22
The K151AKXR-22 is a high-performance thermoelectric module from Mesoscale It has a maximum operating temperature of 150 C and can provide up to 22W of cooling power The module uses bismuth telluride semiconductor material and has a thermal resistance of 0 55 K/W It measures 40 x 40 x 3 4mm in size The K151AKXR-22 requires a heat sink and fan for proper operation With its compact size and solid state operation this thermoelectric module is well-suited for applications such as laser diode cooling medical diagnostics and analytical instrumentation