Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery R-PLEX Human Osteoactivin Assay, singleplex immunoassay, 96-well SECTOR plates, LLOD 12 pg/mL, Goat polyclonal antibodies, For research use only
SDP

Catalog No. NC3169588
Encompass

The R-PLEX Human Osteoactivin Assay is a highly sensitive electrochemiluminescence immunoassay, specifically designed to detect human Osteoactivin. This assay provides comprehensive components essential for robust immunoassay development, enabling effective biomarker discovery and development. It is compatible with various MSD instruments, facilitating quick measurement in small-volume samples. This product is exclusively intended for research applications.

  • Analyte: Osteoactivin
  • Species reactivity: Human
  • Assay format: Singleplex immunoassay
  • Technology: Electrochemiluminescence (ECL)
  • Plate type: 96-well SECTOR plates
  • Lower limit of detection (LLOD): 12 pg/mL
  • Application: Oncology & Cancer
  • Capture antibody: Goat polyclonal
  • Detection antibody: Goat polyclonal
  • Storage condition: Multi-Component
  • Usage: For research use only

Catalog No. NC3169588 Supplier Meso Scale Discovery Supplier No. K151ZHR2
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The R-PLEX Human Osteoactivin Assay is a highly sensitive electrochemiluminescence immunoassay, specifically designed to detect human Osteoactivin. This assay provides comprehensive components essential for robust immunoassay development, enabling effective biomarker discovery and development. It is compatible with various MSD instruments, facilitating quick measurement in small-volume samples. This product is exclusively intended for research applications.

  • Analyte: Osteoactivin
  • Species reactivity: Human
  • Assay format: Singleplex immunoassay
  • Technology: Electrochemiluminescence (ECL)
  • Plate type: 96-well SECTOR plates
  • Lower limit of detection (LLOD): 12 pg/mL
  • Application: Oncology & Cancer
  • Capture antibody: Goat polyclonal
  • Detection antibody: Goat polyclonal
  • Storage condition: Multi-Component
  • Usage: For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.