Learn More
Meso Scale Discovery RPLEX Mouse HAVCR1/KIM1 ASYQP
SDP
Catalog No. 5000081322
Encompass
The R-PLEX Mouse Havcr1 (KIM-1) Assay by Meso Scale Diagnostics measures Havcr1 (KIM-1) in mouse serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity a 5-log dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies and calibrator solutions for a standard curve The assay takes about 3 hours and is used to analyze Havcr1 as a biomarker of kidney toxicity and injury
Catalog No. 50-000-81322 Supplier Meso Scale Discovery Supplier No. K1524UR22
The R-PLEX Mouse Havcr1 (KIM-1) Assay by Meso Scale Diagnostics measures Havcr1 (KIM-1) in mouse serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity a 5-log dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies and calibrator solutions for a standard curve The assay takes about 3 hours and is used to analyze Havcr1 as a biomarker of kidney toxicity and injury