Learn More
Meso Scale Discovery RPLEX Mouse MMP3 total ASYQP
SDP
Catalog No. 5000082086
Encompass
The Meso Scale Discovery R-PLEX Mouse MMP-3 Total Assay is a multiplex immunoassay panel that measures total MMP-3 in mouse biological samples such as serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with low sample volume requirements The assay can be used to monitor total MMP-3 levels in mouse models of disease and pharmacological studies The panel allows flexible customization and validation for additional analytes
Catalog No. 50-000-82086 Supplier Meso Scale Discovery Supplier No. K152ZFR22
The Meso Scale Discovery R-PLEX Mouse MMP-3 Total Assay is a multiplex immunoassay panel that measures total MMP-3 in mouse biological samples such as serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with low sample volume requirements The assay can be used to monitor total MMP-3 levels in mouse models of disease and pharmacological studies The panel allows flexible customization and validation for additional analytes