Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery R-PLEX Mouse Osteoactivin Assay, K152ZHR-2, Singleplex, 96-well, 88 pg/mL LLOD, Dynamic Range: 88-750000 pg/mL, Plasma, Serum
SDP

Catalog No. NC3584827
Encompass

The R-PLEX Mouse Osteoactivin Assay offers researchers a sensitive and efficient way to measure mouse osteoactivin levels in various samples. This assay is tailored for oncological research and is packaged for ease of use. Key features include high sensitivity and a broad dynamic range for effective biomarker profiling.

  • Analyte: Osteoactivin
  • Species: Mouse
  • Assay type: R-PLEX Assay
  • Assay format: Singleplex
  • Application: Oncology & Cancer
  • Plate type: 96-well
  • Sensitivity (LLOD): 88 pg/mL
  • Dynamic range: 88 - 750,000 pg/mL
  • Sample types: Plasma, Serum
  • Research use only: True
  • Storage condition: Multi-Component
  • Gene ID: 93695
  • UniProt ID: Q99P91
  • Kit contents: Mouse Osteoactivin Antibody Set, Mouse Osteoactivin Calibrator
  • Compatible instruments: MESO SECTOR S 600MM, MESO QuickPlex SQ 120MM

Catalog No. NC3584827 Supplier Meso Scale Discovery Supplier No. K152ZHR2
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The R-PLEX Mouse Osteoactivin Assay offers researchers a sensitive and efficient way to measure mouse osteoactivin levels in various samples. This assay is tailored for oncological research and is packaged for ease of use. Key features include high sensitivity and a broad dynamic range for effective biomarker profiling.

  • Analyte: Osteoactivin
  • Species: Mouse
  • Assay type: R-PLEX Assay
  • Assay format: Singleplex
  • Application: Oncology & Cancer
  • Plate type: 96-well
  • Sensitivity (LLOD): 88 pg/mL
  • Dynamic range: 88 - 750,000 pg/mL
  • Sample types: Plasma, Serum
  • Research use only: True
  • Storage condition: Multi-Component
  • Gene ID: 93695
  • UniProt ID: Q99P91
  • Kit contents: Mouse Osteoactivin Antibody Set, Mouse Osteoactivin Calibrator
  • Compatible instruments: MESO SECTOR S 600MM, MESO QuickPlex SQ 120MM

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.