Learn More
Meso Scale Discovery RPLEX Mouse Periostin/OSF2 ASY
SDP
Catalog No. 5000081354
Encompass
The R-PLEX Mouse Periostin (OSF-2) Assay from Meso Scale Discovery is a multiplex immunoassay that measures mouse periostin (OSF-2) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay allows for the quantitative measurement of periostin levels with as little as 25 L of sample It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and a vial of calibrator
Catalog No. 50-000-81354 Supplier Meso Scale Discovery Supplier No. K1529WR2
The R-PLEX Mouse Periostin (OSF-2) Assay from Meso Scale Discovery is a multiplex immunoassay that measures mouse periostin (OSF-2) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay allows for the quantitative measurement of periostin levels with as little as 25 L of sample It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and a vial of calibrator