Learn More
Meso Scale Discovery RPLEX MSE Osteoprotegerin ASY
SDP
Catalog No. 5000078308
Encompass
The Meso Scale Discovery R-PLEX Mouse Osteoprotegerin Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of mouse osteoprotegerin (OPG) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Imagers The assay has a lower limit of detection of 0 04 ng/mL for mouse OPG and a typical assay time of 3 5 hours This assay enables researchers to accurately quantify circulating OPG levels in mouse models
Catalog No. 50-000-78308 Supplier Meso Scale Discovery Supplier No. K152U3R2
The Meso Scale Discovery R-PLEX Mouse Osteoprotegerin Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of mouse osteoprotegerin (OPG) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Imagers The assay has a lower limit of detection of 0 04 ng/mL for mouse OPG and a typical assay time of 3 5 hours This assay enables researchers to accurately quantify circulating OPG levels in mouse models