Learn More
Meso Scale Discovery RPLEX MSE Periostin/OSF2 ASYQP
SDP
Catalog No. 5000081492
Encompass
The R-PLEX Mouse Periostin (OSF-2) Assay from Meso Scale Discovery is a multiplex immunoassay that measures periostin (OSF-2) in mouse samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence (ECL) detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The assay can be fully automated and has a rapid 2 5 hour turnaround time It is available as a singleplex assay that measures only periostin or as a multiplex assay that measures periostin along with other analytes This assay allows the quantitative measurement of periostin which plays a role in cardiac remodeling and bone metabolism
Catalog No. 50-000-81492 Supplier Meso Scale Discovery Supplier No. K1529WR22
The R-PLEX Mouse Periostin (OSF-2) Assay from Meso Scale Discovery is a multiplex immunoassay that measures periostin (OSF-2) in mouse samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence (ECL) detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The assay can be fully automated and has a rapid 2 5 hour turnaround time It is available as a singleplex assay that measures only periostin or as a multiplex assay that measures periostin along with other analytes This assay allows the quantitative measurement of periostin which plays a role in cardiac remodeling and bone metabolism