Learn More
Meso Scale Discovery RPLEX NHP Alphaamylas 2A ASYQP
SDP
Catalog No. 5000078390
Encompass
The R-PLEX NHP Alpha Amylase 2A Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring alpha amylase 2A in human serum plasma saliva and other samples It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and rapid results The kit includes a 96-well plate detection antibody diluents calibrators and controls It quantifies alpha amylase 2A levels between 0 3-1000 ng/mL in 3 5 hours
Catalog No. 50-000-78390 Supplier Meso Scale Discovery Supplier No. K1513IR22
The R-PLEX NHP Alpha Amylase 2A Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring alpha amylase 2A in human serum plasma saliva and other samples It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and rapid results The kit includes a 96-well plate detection antibody diluents calibrators and controls It quantifies alpha amylase 2A levels between 0 3-1000 ng/mL in 3 5 hours