Learn More
Meso Scale Discovery RPLEX NHP Alphaamylase 2A ASY
SDP
Catalog No. 5000078850
Encompass
The R-Plex NHP Alpha-Amylase 2A Assay Kit from Meso Scale Discovery is a 96-well electrochemiluminescent immunoassay for the quantitative measurement of alpha-amylase 2A in human serum plasma saliva and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and has a typical sensitivity of 0 04 ng/mL with an assay range of 0 1-400 ng/mL The kit includes a 96-well MULTI-ARRAY Human Alpha-Amylase 2A Plate SULFO-TAG Anti-Alpha-Amylase 2A Antibody Sample Diluent Read Buffer T (4X) and validations packet This assay allows for high-throughput sensitive and accurate quantification of alpha-amylase 2A levels
Catalog No. 50-000-78850 Supplier Meso Scale Discovery Supplier No. K1513IR2
The R-Plex NHP Alpha-Amylase 2A Assay Kit from Meso Scale Discovery is a 96-well electrochemiluminescent immunoassay for the quantitative measurement of alpha-amylase 2A in human serum plasma saliva and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and has a typical sensitivity of 0 04 ng/mL with an assay range of 0 1-400 ng/mL The kit includes a 96-well MULTI-ARRAY Human Alpha-Amylase 2A Plate SULFO-TAG Anti-Alpha-Amylase 2A Antibody Sample Diluent Read Buffer T (4X) and validations packet This assay allows for high-throughput sensitive and accurate quantification of alpha-amylase 2A levels