Learn More
Novus Biologicals™ rpmB/L28 Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 946941 |
| Species | E. coli |
| Concentration | 0.2 mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP3-23514. |
| Gene Alias | 50 S ribosomal subunit protein L28, ECK3627, Large ribosomal subunit protein bL28, rpmB |
| Gene Symbol | rpmB |
| Quantity | 0.05 mg |
| Storage Buffer | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.