Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci rpmB/L28 Peptide
SDP

Catalog No. 16918910
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-918-910 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-918-910 Supplier ProSci Supplier No. 10061P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

The peptide is used for blocking the activity of the rpmB/L28 antibody.

Specifications

Gene ID (Entrez) 946941
Species E. coli
Accession Number P0A7M2.2
Common Name 50 S ribosomal subunit protein L28
Concentration 200 μg/mL
Content And Storage Store rpmB/L28 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol rpmB
Quantity 0.05 mg
Sequence 15 amino acids amino terminus of E. coli rpmB/L28.
Synonym Large ribosomal subunit protein bL28, 50 S ribosomal protein L28, rpmB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.