Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci rpmH/L34 Peptide
SDP

Catalog No. 16917906
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-917-906 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-917-906 Supplier ProSci Supplier No. 10065P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

The peptide is used for blocking the activity of the rpmH/L34 antibody.

Specifications

Gene ID (Entrez) 948216
Species E. coli
Accession Number P0A7P5.1
Common Name 50 S ribosomal subunit protein L34
Concentration 200 μg/mL
Content And Storage Store rpmH/L34 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol rpmH
Quantity 0.05 mg
Sequence 15 amino acids carboxy terminus of E. coli rpmH/L34.
Synonym rimA, ssaF, Large ribosomal subunit protein bL34, 50 S ribosomal protein L34
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.