Learn More
Sigma Aldrich Fine Chemicals Biosciences RPMI-1640 Medium with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture | MFCD00217820 | 1000ML
Catalog No. NC1748094
Encompass
RPMI-1640 Medium with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture | MFCD00217820 | 1000ML
Catalog No. NC1748094 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 51536C1000ML
RPMI-1640 Medium with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture | MFCD00217820 | 1000ML