Learn More
ATHENA ES RPMI 1640
Catalog No. 5000158176
Encompass_Preferred
Roswell Park Memorial Institute (RPMI) 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins, including certain vitamins not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium.
Catalog No. 50-001-58176 Supplier ATHENA ES Supplier No. 0462
Roswell Park Memorial Institute (RPMI) 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins, including certain vitamins not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium.