Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ RPMI 1640, 1X
BIOPHARMAGSA_VA

Catalog No. MT10104CV Shop All Corning Products
Change view
Click to view available options
Quantity:
6 x 500 mL
Product Type:
RPMI 1640
pH:
7
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type pH
MT10104CV 6 x 500 mL RPMI 1640 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MT10104CV Supplier Corning™ Supplier No. 10104CV
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glutagro™ supplement provides greatly improved stability over the L-glutamine media supplement

Named for the site at which it was developed, the Roswell Park Memorial Institute, RPMI 1640 is a modification of McCoy's 5A containing an increased level of inositol. Cell types for which this medium is appropriate include mammalian myeloma and hybridoma cells such as mouse hybridomas, human leukocytes, and B and T lymphocytes.

  • Can be used for suspension cultures or monolayer human leukemic cells
  • This formulation is supplemented with glutaGRO™ in place of L-glutamine for improved stability

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type RPMI 1640
Sterility Sterile
With Additives glutagro Supplement, Phenol Red
Quantity 6 x 500 mL
pH 7
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.