Learn More
Sigma Aldrich Fine Chemicals Biosciences RPMI 1640 Complete Medium, with 2mM L-Glut and 10% FBS, 500 mL
Catalog No. NC1691620
Encompass
RPMI 1640 Complete Medium is supplemented with 2mM L-Glutamine and 10% U.S. origin FBS. It is a convenient sterile solution for the culture of a wide variety of cells that grow in suspension or as anchorage dependent, and can be used in fusion protocols and in the growth of hybrid cells.
Catalog No. NC1691620 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SLM240B
RPMI 1640 Complete Medium is supplemented with 2mM L-Glutamine and 10% U.S. origin FBS. It is a convenient sterile solution for the culture of a wide variety of cells that grow in suspension or as anchorage dependent, and can be used in fusion protocols and in the growth of hybrid cells.